Anti-WASF1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: S91-36]
Proveedor: Abnova
MAB11594
ABNOMAB11594EA 545 EUR
ABNOMAB11594
Anti-WASF1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: S91-36]
Anticuerpos
Mouse monoclonal antibody raised against synthetic peptide of WASF1.
Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Western Blot: 1 ?g/ml
Type: Primary
Antigen: WASF1
Clonality: Monoclonal
Clone: S91-36
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now