Anti-KCNB1 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: S89-34]

Proveedor: Abnova
ABNOMAB11582EA 545 EUR
ABNOMAB11582
Anti-KCNB1 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: S89-34]
Anticuerpos
Mouse monoclonal antibody raised against synthetic peptide of Kcnb1.

Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Western Blot: 1 ?g/ml

Type: Primary
Antigen: KCNB1
Clonality: Monoclonal
Clone: S89-34
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR