Anti-KCNB1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: S89-34]
Proveedor: Abnova
MAB11576
ABNOMAB11576EA 545 EUR
ABNOMAB11576
Anti-KCNB1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: S89-34]
Anticuerpos
Mouse monoclonal antibody raised against synthetic peptide of Kcnb1.
Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Western Blot: 1 ?g/ml
Type: Primary
Antigen: KCNB1
Clonality: Monoclonal
Clone: S89-34
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now