Anti-CK4 Mouse Polyclonal Antibody

Proveedor: Abnova
ABNOH00003851-B01PEA 380 EUR
ABNOH00003851-B01P
Anti-CK4 Mouse Polyclonal Antibody
Anticuerpos
Mouse polyclonal antibody raised against a full-length human KRT4 protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: CK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR