Anti-IGFBP5 Mouse Polyclonal Antibody

Proveedor: Abnova
ABNOH00003488-B01PEA 380 EUR
ABNOH00003488-B01P
Anti-IGFBP5 Mouse Polyclonal Antibody
Anticuerpos
Mouse polyclonal antibody raised against a full-length human IGFBP5 protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: IGFBP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR