Anti-CDX2 Mouse Monoclonal Antibody [clone: 1C19]
Proveedor: Abnova
H00001045-M02
ABNOH00001045-M02EA 394 EUR
ABNOH00001045-M02
Anti-CDX2 Mouse Monoclonal Antibody [clone: 1C19]
Anticuerpos
Mouse monoclonal antibody raised against a full-length recombinant CDX2.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: CDX2
Clonality: Monoclonal
Clone: 1C19
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity:
Order Now