Anti-c-Myc tag Chicken Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Proveedor: Abnova
PAB29203
ABNOPAB29203EA 350 EUR
ABNOPAB29203
Anti-c-Myc tag Chicken Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Anticuerpos
Chicken polyclonal antibody raised against c-Myc tag with FITC conjugation.
Recommended Dilutions: Western Blot: 1:500
Type: Tag
Antigen: c-Myc tag
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now