Anti-TG Chicken Polyclonal Antibody (Biotin)

Proveedor: Abnova
ABNOPAB29199EA 356 EUR
ABNOPAB29199
Anti-TG Chicken Polyclonal Antibody (Biotin)
Anticuerpos
Chicken polyclonal antibody raised against native Human TG with Biotin conjugation.

Type:
Antigen: TG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR