Anti-TG Chicken Polyclonal Antibody

Proveedor: Abnova
ABNOPAB29198EA 356 EUR
ABNOPAB29198
Anti-TG Chicken Polyclonal Antibody
Anticuerpos
Chicken polyclonal antibody raised against native Human TG.

Type: Primary
Antigen: TG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR