Anti-ALB Chicken Polyclonal Antibody

Proveedor: Abnova
ABNOPAB29197EA 338 EUR
ABNOPAB29197
Anti-ALB Chicken Polyclonal Antibody
Anticuerpos
Chicken polyclonal antibody raised against native Human ALB.

Type: Primary
Antigen: ALB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR