Anti-PECAM1 Mouse Monoclonal Antibody [clone: SPM532]
Proveedor: Abnova
MAB12123
ABNOMAB12123EA 518 EUR
ABNOMAB12123
Anti-PECAM1 Mouse Monoclonal Antibody [clone: SPM532]
Anticuerpos
Mouse monoclonal antibody raised against PECAM1.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: PECAM1
Clonality: Monoclonal
Clone: SPM532
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now