Anti-PCNA Mouse Monoclonal Antibody [clone: SPM350]
Proveedor: Abnova
ABNOMAB12122EA 440 EUR
ABNOMAB12122
Anti-PCNA Mouse Monoclonal Antibody [clone: SPM350]
Anticuerpos
Mouse monoclonal antibody raised against PCNA.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: PCNA
Clonality: Monoclonal
Clone: SPM350
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now