Anti-ACTA1 Mouse Monoclonal Antibody [clone: SPM160]
Proveedor: Abnova
ABNOMAB12070EA 440 EUR
ABNOMAB12070
Anti-ACTA1 Mouse Monoclonal Antibody [clone: SPM160]
Anticuerpos
Mouse monoclonal antibody raised against muscle specific actin.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: ACTA1
Clonality: Monoclonal
Clone: SPM160
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now