Anti-TP53 Mouse Monoclonal Antibody [clone: SPM590]
Proveedor: Abnova
ABNOMAB12060EA 440 EUR
ABNOMAB12060
Anti-TP53 Mouse Monoclonal Antibody [clone: SPM590]
Anticuerpos
Mouse monoclonal antibody raised against recombinant TP53.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: SPM590
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Order Now