Anti-SLC17A7 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: S28-9]
Proveedor: Abnova
ABNOMAB11521EA 643 EUR
ABNOMAB11521
Anti-SLC17A7 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: S28-9]
Anticuerpos
Mouse monoclonal antibody raised against recombinant of Slc17a7.
Recommended Dilutions: Immunohistochemistry Western Blot: 1:1000
Type: Primary
Antigen: SLC17A7
Clonality: Monoclonal
Clone: S28-9
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now