Anti-SLC5A5 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: FP5]

Proveedor: Abnova
MAB11503
ABNOMAB11503EA 643 EUR
ABNOMAB11503
Anti-SLC5A5 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: FP5]
Anticuerpos
Mouse monoclonal antibody raised against recombinant of SLC5A5.

Recommended Dilutions: Immunofluorescence Immunohistochemistry Western Blot: 1:1000

Type: Primary
Antigen: SLC5A5
Clonality: Monoclonal
Clone: FP5
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR