Anti-SLC5A5 Mouse Monoclonal Antibody [clone: FP5]
Proveedor: Abnova
ABNOMAB11494EA 440 EUR
ABNOMAB11494
Anti-SLC5A5 Mouse Monoclonal Antibody [clone: FP5]
Anticuerpos
Mouse monoclonal antibody raised against recombinant of SLC5A5.
Recommended Dilutions: Immunofluorescence Immunohistochemistry Western Blot: 1:1000
Type: Primary
Antigen: SLC5A5
Clonality: Monoclonal
Clone: FP5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now