Anti-FASLG Mouse Polyclonal Antibody
Proveedor: Abnova
ABNOH00000356-B01PEA 380 EUR
ABNOH00000356-B01P
Anti-FASLG Mouse Polyclonal Antibody
Anticuerpos
Mouse polyclonal antibody raised against a full-length human FASLG protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: FASLG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now