Anti-MLANA Mouse Monoclonal Antibody [clone: SPM342]

Proveedor: Abnova
ABNOMAB12086EA 440 EUR
ABNOMAB12086
Anti-MLANA Mouse Monoclonal Antibody [clone: SPM342]
Anticuerpos
Mouse monoclonal antibody raised against recombinant MLANA.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: MLANA
Clonality: Monoclonal
Clone: SPM342
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR