Anti-TAG72 Mouse Monoclonal Antibody [clone: SPM536]

Proveedor: Abnova
ABNOMAB12083EA 440 EUR
ABNOMAB12083
Anti-TAG72 Mouse Monoclonal Antibody [clone: SPM536]
Anticuerpos
Mouse monoclonal antibody raised against tag-72.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: TAG72
Clonality: Monoclonal
Clone: SPM536
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR