Anti-Granulocyte Mouse Monoclonal Antibody [clone: SPM250]

Proveedor: Abnova
MAB12080
ABNOMAB12080EA 518 EUR
ABNOMAB12080
Anti-Granulocyte Mouse Monoclonal Antibody [clone: SPM250]
Anticuerpos
Mouse monoclonal antibody raised against Granulocyte.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: Granulocyte
Clonality: Monoclonal
Clone: SPM250
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR