Anti-EPCAM Mouse Monoclonal Antibody [clone: SPM535]

Proveedor: Abnova
MAB12079
ABNOMAB12079EA 518 EUR
ABNOMAB12079
Anti-EPCAM Mouse Monoclonal Antibody [clone: SPM535]
Anticuerpos
Mouse monoclonal antibody raised against Tacstd1.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: EPCAM
Clonality: Monoclonal
Clone: SPM535
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR