Anti-FN1 Chicken Polyclonal Antibody

Proveedor: Abnova
PAB29174
ABNOPAB29174EA 338 EUR
ABNOPAB29174
Anti-FN1 Chicken Polyclonal Antibody
Anticuerpos
Chicken polyclonal antibody raised against native Human FN1.

Recommended Dilutions: ELISA: 1:10000

Type: Primary
Antigen: FN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR