Anti-IgY F(c) Monoclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: Abnova
ABNOPAB29164EA 229 EUR
ABNOPAB29164
Anti-IgY F(c) Monoclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Donkey anti-chicken IgY recognizes chicken IgY F(c). The anti-F(c)-specific fraction was purified using antigen affinity chromatography.

Type: Secondary
Antigen: IgY F(c)
Clonality: Monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: F(c)
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR