Anti- IgY F(c) Monoclonal Antibody (Unconjugated)

Proveedor: Abnova
ABNOPAB29161EA 170 EUR
ABNOPAB29161
Anti- IgY F(c) Monoclonal Antibody (Unconjugated)
Anticuerpos
Donkey anti-chicken IgY recognizes chicken IgY F(c). The anti-F(c)-specific fraction was purified using antigen affinity chromatography.

Type: Secondary
Antigen: IgY F(c)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR