Anti-Clostridium difficile Toxin A Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))
Proveedor: Abnova
ABNOPAB29153EA 440 EUR
ABNOPAB29153
Anti-Clostridium difficile Toxin A Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Chicken polyclonal antibody raised against Clostridium difficile Toxin A.
Type:
Antigen: Clostridium difficile Toxin A
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now