Anti-CHGA Mouse Monoclonal Antibody [clone: SPM585]
Proveedor: Abnova
ABNOMAB12042EA 518 EUR
ABNOMAB12042
Anti-CHGA Mouse Monoclonal Antibody [clone: SPM585]
Anticuerpos
Mouse monoclonal antibody raised against CHGA.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: CHGA
Clonality: Monoclonal
Clone: SPM585
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now