Anti-CEACAM5 Mouse Monoclonal Antibody [clone: SPM541]
Proveedor: Abnova
ABNOMAB12037EA 440 EUR
ABNOMAB12037
Anti-CEACAM5 Mouse Monoclonal Antibody [clone: SPM541]
Anticuerpos
Mouse monoclonal antibody raised against CEACAM5.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: CEACAM5
Clonality: Monoclonal
Clone: SPM541
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now