Anti-CD79A Mouse Monoclonal Antibody [clone: SPM550]
Proveedor: Abnova
ABNOMAB12034EA 440 EUR
ABNOMAB12034
Anti-CD79A Mouse Monoclonal Antibody [clone: SPM550]
Anticuerpos
Mouse monoclonal antibody raised against synthetic peptide of CD79A.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: CD79A
Clonality: Monoclonal
Clone: SPM550
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now