Anti-CD79A Mouse Monoclonal Antibody [clone: SPM549]

Proveedor: Abnova
ABNOMAB12033EA 440 EUR
ABNOMAB12033
Anti-CD79A Mouse Monoclonal Antibody [clone: SPM549]
Anticuerpos
Mouse monoclonal antibody raised against synthetic peptide of CD79A.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: CD79A
Clonality: Monoclonal
Clone: SPM549
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR