Anti-CD34 Mouse Monoclonal Antibody [clone: SPM123]

Proveedor: Abnova
MAB12030
ABNOMAB12030EA 518 EUR
ABNOMAB12030
Anti-CD34 Mouse Monoclonal Antibody [clone: SPM123]
Anticuerpos
Mouse monoclonal antibody raised against CD34.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: SPM123
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR