Anti-CA9 Mouse Monoclonal Antibody [clone: SPM314]

Proveedor: Abnova
MAB12024
ABNOMAB12024EA 518 EUR
ABNOMAB12024
Anti-CA9 Mouse Monoclonal Antibody [clone: SPM314]
Anticuerpos
Mouse monoclonal antibody raised against CA9.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: CA9
Clonality: Monoclonal
Clone: SPM314
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR