Anti-AFP Mouse Monoclonal Antibody [clone: SPM334]

Proveedor: Abnova
ABNOMAB12017EA 518 EUR
ABNOMAB12017
Anti-AFP Mouse Monoclonal Antibody [clone: SPM334]
Anticuerpos
Mouse monoclonal antibody raised against AFP.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: SPM334
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR