Anti-ATX Chicken Polyclonal Antibody

Proveedor: Abnova
ABNOPAB29131EA 545 EUR
ABNOPAB29131
Anti-ATX Chicken Polyclonal Antibody
Anticuerpos
Chicken polyclonal antibody raised against Clostridium botulinum Type A Neurotoxin Heavy Chain.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:2000

Type: Primary
Antigen: ATX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR