Anti-BSA Chicken Polyclonal Antibody

Proveedor: Abnova
ABNOPAB29123EA 399 EUR
ABNOPAB29123
Anti-BSA Chicken Polyclonal Antibody
Anticuerpos
Chicken polyclonal antibody raised against native BSA.

Recommended Dilutions: ELISA: 1:200-1:500 for capture; 1:1000-1:20000 for primary antibody)

Type: Primary
Antigen: BSA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR