Anti-IgG Chicken Monoclonal Antibody (HRP (Horseradish Peroxidase))
Proveedor: Abnova
ABNOPAB29110EA 223 EUR
ABNOPAB29110
Anti-IgG Chicken Monoclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Chicken polyclonal antibody raised against Rabbit IgG. The antibody is conjugated with HRP.
Recommended Dilutions: Western Blot: 1:5000
Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Rabbit