Anti-SCN8A Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: S87A-10]
Proveedor: Abnova
ABNOMAB11995EA 545 EUR
ABNOMAB11995
Anti-SCN8A Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: S87A-10]
Anticuerpos
Mouse monoclonal antibody raised against synthetic peptide of Scn8a.
Recommended Dilutions: Immunocytochemistry (0.1-1 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml
Type: Primary
Antigen: SCN8A
Clonality: Monoclonal
Clone: S87A-10
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now