Anti-SCN8A Mouse Monoclonal Antibody (ATTO 390) [clone: S87A-10]

Proveedor: Abnova
MAB11981
ABNOMAB11981EA 545 EUR
ABNOMAB11981
Anti-SCN8A Mouse Monoclonal Antibody (ATTO 390) [clone: S87A-10]
Anticuerpos
Mouse monoclonal antibody raised against synthetic peptide of Scn8a.

Recommended Dilutions: Immunocytochemistry (0.1-1 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml

Type: Primary
Antigen: SCN8A
Clonality: Monoclonal
Clone: S87A-10
Conjugation: ATTO 390
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR