Anti-SCN1A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: S74-71]
Proveedor: Abnova
MAB11972
ABNOMAB11972EA 545 EUR
ABNOMAB11972
Anti-SCN1A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: S74-71]
Anticuerpos
Mouse monoclonal antibody raised against partial recombinant Scn1a.
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml
Type: Primary
Antigen: SCN1A
Clonality: Monoclonal
Clone: S74-71
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now