Anti-MAPT Chicken Polyclonal Antibody

Proveedor: Abnova
ABNOPAB29092EA 655 EUR
ABNOPAB29092
Anti-MAPT Chicken Polyclonal Antibody
Anticuerpos
Chicken polyclonal antibody raised against recombinant Human MAPT.

Recommended Dilutions: Immunocytochemistry: 1:1000-1:2000; Immunohistochemistry: 1:1000-1:2000; Western Blot

Type: Primary
Antigen: MAPT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR