Anti-SYT1 Chicken Polyclonal Antibody

Proveedor: Abnova
ABNOPAB29091EA 655 EUR
ABNOPAB29091
Anti-SYT1 Chicken Polyclonal Antibody
Anticuerpos
Chicken polyclonal antibody raised against recombinant Human SYT1.

Recommended Dilutions: Immunocytochemistry: 1:1000-1:2000; Immunohistochemistry: 1:1000-1:2000; Western Blot: 1:2000-1:5000

Type: Primary
Antigen: SYT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR