Anti-PLP1 Chicken Polyclonal Antibody

Proveedor: Abnova
ABNOPAB29088EA 772 EUR
ABNOPAB29088
Anti-PLP1 Chicken Polyclonal Antibody
Anticuerpos
Chicken polyclonal antibody raised against recombinant Human PLP1.

Recommended Dilutions: Immunocytochemistry: 1:1000-1:2000; Immunohistochemistry: 1:1000-1:2000

Type: Primary
Antigen: PLP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR