Anti-ACPP Chicken Polyclonal Antibody

Proveedor: Abnova
ABNOPAB29086EA 356 EUR
ABNOPAB29086
Anti-ACPP Chicken Polyclonal Antibody
Anticuerpos
Chicken polyclonal antibody raised against recombinant Mouse Acpp.

Recommended Dilutions: Immunocytochemistry Immunohistochemistry: 1:500-1:1000

Type: Primary
Antigen: ACPP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR