Anti-NEFH Chicken Polyclonal Antibody

Proveedor: Abnova
ABNOPAB29081EA 356 EUR
ABNOPAB29081
Anti-NEFH Chicken Polyclonal Antibody
Anticuerpos
Chicken polyclonal antibody raised against recombinant Bovine NEFH.

Recommended Dilutions: Immunocytochemistry: 1:2000-1:5000; Immunohistochemistry: 1:2000-1:5000; Western Blot: 1:5000-1:10000

Type: Primary
Antigen: NEFH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR