Anti-NES Chicken Polyclonal Antibody

Proveedor: Abnova
ABNOPAB29079EA 772 EUR
ABNOPAB29079
Anti-NES Chicken Polyclonal Antibody
Anticuerpos
Chicken polyclonal antibody raised against recombinant Mouse Nes.

Recommended Dilutions: Immunocytochemistry: 1:10000-1:20000; Immunohistochemistry: 1:10000-1:20000; Western Blot: 1:20000-1:50000

Type: Primary
Antigen: NES
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR