Anti-SCN1A Mouse Monoclonal Antibody (ATTO 390) [clone: S74-71]

Proveedor: Abnova
ABNOMAB11963EA 545 EUR
ABNOMAB11963
Anti-SCN1A Mouse Monoclonal Antibody (ATTO 390) [clone: S74-71]
Anticuerpos
Mouse monoclonal antibody raised against partial recombinant Scn1a.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml

Type: Primary
Antigen: SCN1A
Clonality: Monoclonal
Clone: S74-71
Conjugation: ATTO 390
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR