Anti-GRM3 Chicken Polyclonal Antibody
Proveedor: Abnova
ABNOPAB29071EA 568 EUR
ABNOPAB29071
Anti-GRM3 Chicken Polyclonal Antibody
Anticuerpos
Chicken polyclonal antibody raised against recombinant Human GRM3.
Recommended Dilutions: Immunocytochemistry: 1:1000-1:2000; Immunohistochemistry: 1:1000-1:2000
Type: Primary
Antigen: GRM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now