Anti-GRM2 Chicken Polyclonal Antibody

Proveedor: Abnova
ABNOPAB29070EA 568 EUR
ABNOPAB29070
Anti-GRM2 Chicken Polyclonal Antibody
Anticuerpos
Chicken polyclonal antibody raised against recombinant Human GRM2.

Recommended Dilutions: Immunocytochemistry: 1:1000-1:2000; Immunohistochemistry: 1:1000-1:2000

Type: Primary
Antigen: GRM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR