Anti-c-Myc Polyclonal Antibody

Proveedor: Abnova
ABNOPAB29060EA 518 EUR
ABNOPAB29060
Anti-c-Myc Polyclonal Antibody
Anticuerpos
Chicken polyclonal antibody raised against recombinant c-Myc peptide.

Recommended Dilutions: Immunocytochemistry: 1:250; Immunoprecipitation: 1:250; Western Blot: 1:500

Type: Primary
Antigen: c-Myc
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR