Anti-GLB1 Polyclonal Antibody

Proveedor: Abnova
ABNOPAB29055EA 338 EUR
ABNOPAB29055
Anti-GLB1 Polyclonal Antibody
Anticuerpos
Chicken polyclonal antibody raised against purified beta-galactosidase.

Recommended Dilutions: Immunocytochemistry: 1:2500-1:5000; Immunohistochemistry: 1:2500-1:5000; Western Blot: 1:5000-1:10000

Type: Primary
Antigen: GLB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR